Upchurch Scientific™ Low-Pressure Unions: PEEK DFS Item
Manufacturer:  Upchurch Scientific™ UPP705
 View more versions of this product
Catalog No. 05-700-174
Specifications
Specifications
Union
0.02 in.
5000 psi
125°C
Thru Hole: 0.020 in.
PEEK
Black
Documents