Upchurch Scientific™ Low-Pressure Unions: PEEK DFS Item
Manufacturer:  Upchurch Scientific™ UPP703
 View more versions of this product
Catalog No. 05-700-172
Specifications
Specifications
Union
PEEK
Natural
With 2 P-300/P-335, 1/8 in., 1/4-28, 0.050 in. (1.25 mm) THRU
1000 psi
125°C
Documents