Idex Low-Pressure Inline Filters DFS Item
These filters offer excellent protection and allow for tailoring to a variety of low-pressure applications.
Supplier:  Idex UPA416
 View more versions of this product
Catalog No. 05-707-43
Description
Description
Each filter is rated to 1,000 psi (68.9 bar). They work best when used with PTFE or ETFE tubing, but can be used with any appropriate size semi-rigid or rigid tubing.
  • Zero-dead volume connections
  • No wrenches required
  • For 1/16 in. or 1/8 in. OD tubing
When choosing alternate or replacement frits, select only those with an overall diameter of .200 in., including polymer ring.
Specifications
Specifications
2 μm
1/8 in. O.D. tubing
Stainless Steel