Upchurch Scientific™ Low-Pressure Inline Filters
These filters offer excellent protection and allow for tailoring to a variety of low-pressure applications.
$65.63 - $65.63
Specifications
Pore Size 2μm
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Housing Material Price Quantity  
Catalog Number Mfr. No. Housing Material Price Quantity  
05-707-43 Upchurch Scientific™
UPA416
Stainless Steel Each for $65.63
Specifications
Description
Each filter is rated to 1,000 psi (68.9 bar). They work best when used with PTFE or ETFE tubing, but can be used with any appropriate size semi-rigid or rigid tubing.
  • Zero-dead volume connections
  • No wrenches required
  • For 1/16 in. or 1/8 in. OD tubing
When choosing alternate or replacement frits, select only those with an overall diameter of .200 in., including polymer ring.
Description & Specifications
Specifications
2μm