Upchurch Scientific™ Inlet Solvent Filters: End Fittings
Manufactured from Polyether ether ketone (PEEK). Upchurch Scientific Inlet Solvent Filters: End Fittings are used for Analytical HPLC Systems.
$142.00 - $148.67
Specifications
Product Type Replacement Filter End Fitting
Material PEEK
Thread Size 10 to 32 Coned
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Pore Size Price Quantity  
Catalog Number Mfr. No. Pore Size Price Quantity  
05-707-83 Upchurch Scientific™
UPA428X*
0.5μm Pack of 10 Each for $148.67
05-707-84 Upchurch Scientific™
UPA429X*
0.2μm Pack of 10 Each for $142.00
Specifications
Description
  • For Analytical HPLC System
  • 0.5µm or 0.2µm PEEK filter end fittings
  • Black or natural body
  • Swept volume of 5.7µL or 6.9µL
  • Thread port: 10-32 Coned
Description & Specifications
Specifications
Replacement Filter End Fitting
PEEK
10 to 32 Coned