Upchurch Scientific™ Inlet Solvent Filters DFS Item
Stainless-steel or multiple materials
Manufacturer:  Upchurch Scientific™ UPA302A
 View more versions of this product
Catalog No. 05-707-25
Description
Description
Cups and end fittings sold separately, see 05-704-54 and 045-707-83.
Specifications
Specifications
40 mL/min
Stainless Steel
0.25 to 28 Flat bottom
1 Filter (Cups and end fittings sold separately)
10 µm
Inlet Solvent Filter
Documents