Idex High-Pressure Inline Filters DFS Item
Prevent problems stemming from particulate matter and protect major system components by using these filters
Supplier:  Idex UPA314
 View more versions of this product
Catalog No. 29-008-1001
Description
Description
Prevent problems stemming from particulate matter and protect major system components by using these filters
  • Excellent choices for inline filtration needs
  • Multiple versions customized for different applications available
Specifications
Specifications
2 μm
Stainless Steel