Please login to your online account to display your discounted pricing
Upchurch Scientific™ Flangeless Nuts: Compatible with 1/4-28, PFA
$44.89 - $44.89
Specifications
Certifications/Compliance ROHS
Color Natural
Dimensions (L x W) 0.87 x 0.37 in. (2.2 x 9.4cm)
Length (English) Thread 0.48 in.
Material PFA
View More Specs
Products
Catalog Number Mfr. No. For Use With (Equipment) Max. Pressure Product Type Price Quantity    
29-008-530
upchurch scientific™
UPP345X
For 0.13 in. O.D. tubing 500psi Flangeless standard knurled head short nut Pack of 10 Each for $44.89
Description & Specifications
Specifications
Certifications/Compliance ROHS
Color Natural
Dimensions (L x W) 0.87 x 0.37 in. (2.2 x 9.4cm)
Length (English) Thread 0.48 in.
Material PFA
Port Style 0.25 to 28 flat bottom
Quantity 10/Pack