missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Upchurch Scientific™ Flangeless Nuts: Compatible with 1/4-28, PEEK DFS Item

Manufacturer:  Upchurch Scientific™ UPP235X

 View more versions of this product

Catalog No. 05-700-394



Specifications

Specifications

ROHS
0.61 x 0.37 in. (1.54 x 9.4 cm)
0.61 in.
1.54 cm
2000 psi
Flangeless standard knurled head short nut
9.4 cm
For 0.06 in. O.D. Tubing
Natural
0.35 in.
0.89 cm
PEEK
0.25 to 28 flat bottom
0.37 in.
10/Pk.
Documents