missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Upchurch Scientific™ Flangeless Nuts: Compatible with 1/4-28, PEEK DFS Item

Manufacturer:  Upchurch Scientific™ P235

 View more versions of this product

Catalog No. 05-700-393



Specifications

Specifications

ROHS
PEEK
0.25 to 28 flat bottom
Each
Natural
2000 psi
Flangeless standard knurled head short nut
For 0.06 in. O.D. Tubing
Documents