missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Upchurch Scientific™ Flangeless Fittings: Ferrules, PEEK

$73.80 - $73.80

Specifications

Color Natural
Type Flangeless Ferrule
For Use With 1/16 in. O.D. Tubing
Material PEEK
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Length (English) Width (English) Price Quantity  
Catalog Number Mfr. No. Length (English) Width (English) Price Quantity  
05-700-92 Upchurch Scientific™
UPP250X
0.16 in. 0.18 in. Pack of 10 Each for $73.80
Specifications Description & Specifications

Specifications

Natural
Flangeless Ferrule
1/16 in. O.D. Tubing
PEEK