missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Upchurch Scientific™ FEP Tubing

Chemically inert and suitable for lower pressure applications

$85.52 - $85.52

Specifications

Certifications/Compliance ROHS
For Use With (Application) For general, low pressure appliCatIons
Material DuPont™ FEP Fluoropolymer
pH Range 0 to 14
Temperature (English) Maximum 122°F
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Color Length (English) Max. Pressure Pressure Price Quantity  
Catalog Number Mfr. No. Color Length (English) Max. Pressure Pressure Price Quantity  
05-701-48 Upchurch Scientific™
UP1527
Natural 20 ft. 1000 to 3000psi 1000-3000psi Each for $85.52
Specifications

Description

  • Great for general, low pressure applications
  • Features virtually identical chemical inertness when compared with PTFE
  • Some sizes available in multiple, translucent colors for easy flow path identification
  • Some sizes also in black for light sensitive applications

Description & Specifications

Specifications

ROHS
DuPont™ FEP Fluoropolymer
122°F
For general, low pressure appliCatIons
0 to 14
50°C