Idex Bottom-Of-The-Bottle™ Inlet Filters DFS Item
Protect your system from damaging particulates
$79.58 - $251.91
Specifications
Certifications/Compliance ROHS
Filter Type Bottom-of-the-Bottle™
For Use With (Application) Analytical instrumentation, in vitro diagnostics, biotechnology, semiconductor/electronics, laboratory applications
No. per Case Each
Type Inlet Solvent Filter
View More Specs
Products 6
Catalog Number Mfr. No. Material Pore Size Flow Rate For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. Material Pore Size Flow Rate For Use With (Equipment) Price Quantity  
05-707-103 DFS Item
View Documents Promotion Details
Idex
UPA440
PEEK 10 μm 30 mL/min. For 0.05 in. O.D tubing
Each for $83.79
05-707-57 DFS Item
View Documents Promotion Details
Idex
UPA550*
PEEK, ETFE, Stainless Steel 10 μm 40 mL/min. For 0.05 in. I.D tubing
Each for $251.91
05-707-58 DFS Item
View Documents Promotion Details
Idex
UPA551
PEEK, ETFE, Stainless Steel 2 μm 10 mL/min. For 0.05 in. O.D tubing
Each for $229.83
05-707-87 DFS Item
View Documents Promotion Details
Idex
UPA309
Stainless Steel 10 μm 40 mL/min. For 0.0625 in. O.D tubing
Each for $85.79
29-008-971 DFS Item
View Documents Promotion Details
Idex
UPA438
PEEK 10 μm 30 mL/min. For 0.05 in. O.D tubing (small neck)
Each for $85.21
29-008-970 DFS Item
View Documents Promotion Details
Idex
UPA437
PEEK 2 μm 30 mL/min. For 0.05 in. O.D tubing (small neck), w/2 PEEK frits
Each for $79.58
Description
Description
  • Filters are biocompatible—constructed of 100% PEEK polymer
  • Use with 1/8 in. OD tubing
Specifications
Specifications
ROHS
Analytical instrumentation, in vitro diagnostics, biotechnology, semiconductor/electronics, laboratory applications
Inlet Solvent Filter
Bottom-of-the-Bottle™
Each
SDS
missing translation for 'documents'
missing translation for 'documents'