Please login to your online account to display your discounted pricing
Upchurch Scientific™ Barbed Reducers
Manufacturer: upchurch scientific™  UPP807X
 View more versions of this product
Catalog No. 05-700-200
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications
Specifications
Type Barbed Reducing Adapter
For Use With (Equipment) To connect 1/8 in. ID (3.20 mm) Tubing to 1/16 in. ID (1.55 mm) Tubing