Please login to your online account to display your discounted pricing
Upchurch Scientific™ Back Pressure Regulators: Cartridges
For use with Upchurch Scientific Back Pressure Regulators
$149.48 - $159.94
Specifications
Type Cartridge
For Use With (Application) Back Pressure Regulators
View More Specs
Products
Catalog Number Mfr. No. Pressure Swept Volume Price Quantity    
05-704-212
upchurch scientific™
UPP763
100psi 125μL Each for $159.94
05-704-211
upchurch scientific™
P762
75psi 125μL Each for $159.19
29-008-1220
upchurch scientific™
UPP795
750psi 87μL Each for $159.87
05-704-210
upchurch scientific™
UPP761*
40psi 125μL Each for $159.19
05-704-213
upchurch scientific™
UPP764
250psi 95μL Each for $149.48
Description & Specifications
Specifications
Type Cartridge
For Use With (Application) Back Pressure Regulators
Biocompatible/chemically resistant materials.