Idex Adapters for HPLC DFS Item
Convenience of quick connect and disconnect of a luer connection
Manufacturer:  Idex P647
 View more versions of this product
Catalog No. 05-700-315
Description
Description
  • Manufactured from inert polymers
Quick connect luer adapters are designed to connect any female luer to any male luer in a variety of applications. Use with male luer lock adapters for more secure connection.
Specifications
Specifications
Natural
ETFE
0.08 in.
1/8 in. I.D. Tubing
Adapter
Barb to Male Luer Lock
Documents