Thermo Scientific™ uPA Inhibitor II, UK122
Supplier: Thermo Scientific™ J64874MB
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- A cell-permeable oxazolidinone-carboxamidine compound that acts as a potent, selective, and active-site targeting inhibitor of urokinase-type plasminogen activator
Specifications
25 mg | |
2°C to 8°C | |
A cell-permeable oxazolidinone-carboxamidine compound that acts as a potent, selective, and active-site targeting inhibitor of urokinase-type plasminogen activator | |
405.30g/mol |
Yellow | |
Solid | |
C17H13N3O2·CF3CO2H | |
Light Sensitive |
RUO â Research Use Only