missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
unsym-Dimethylethylenediamine, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 116201000
Specifications
unsym-Dimethylethylenediamine | |
12°C | |
Glass bottle | |
0.8 | |
105.5°C to 107°C | |
-70°C | |
108-00-9 | |
C4H12N2 | |
MFCD00008175 | |
1,2-ethanediamine, n,n-dimethyl, 2-aminoethyl dimethylamine, 2-aminoethyldimethylamine, 2-dimethylamino ethylamine, 2-dimethylaminoethylamine, n,n-dimethyl-1,2-ethanediamine, n,n-dimethyl-1,2-ethylenediamine, n,n-dimethylethanediamine, n,n-dimethylethylenediamine, n1,n1-dimethylethane-1,2-diamine | |
DILRJUIACXKSQE-UHFFFAOYSA-N | |
N',N'-dimethylethane-1,2-diamine | |
66053 | |
Liquid |
0.8000g/mL | |
Authentic | |
1.4255 to 1.4275 | |
1% max. | |
Colorless to Yellow | |
100g | |
98.5% min. (GC) | |
(CH3)2NCH2CH2NH2 | |
04, II, 690 | |
Solubility in water: miscible with water | |
CN(C)CCN | |
88.15 | |
88.15 | |
99% |
Chemical Identifiers
108-00-9 | |
88.15 | |
DILRJUIACXKSQE-UHFFFAOYSA-N | |
66053 | |
CN(C)CCN |
C4H12N2 | |
MFCD00008175 | |
1,2-ethanediamine, n,n-dimethyl, 2-aminoethyl dimethylamine, 2-aminoethyldimethylamine, 2-dimethylamino ethylamine, 2-dimethylaminoethylamine, n,n-dimethyl-1,2-ethanediamine, n,n-dimethyl-1,2-ethylenediamine, n,n-dimethylethanediamine, n,n-dimethylethylenediamine, n1,n1-dimethylethane-1,2-diamine | |
N',N'-dimethylethane-1,2-diamine |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Highly flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse
GHS Signal Word: Danger
EINECSNumber : 203-541-2