Please login to your online account to display your discounted pricing

Unleaded gasoline solution (unweathered), 5000μg/mL in methanol, Ultra Scientific

$124.11 - $124.11

Chemical Identifiers

Chemical Name or Material Unleaded gasoline solution (unweathered)
CAS 000067-56-1
CAS Max % 99.37
CAS 008006-61-9
CAS Max % 0.63
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

US-RGO-601-1

 
ultra scientific
RGO6011
1 x 1mL Each for $124.11
Description & Specifications

Specifications

Quantity 1 x 1mL
Solvent or Matrix Methanol (Methyl alcohol)
Analyte or Component Names Methanol,gasoline (regular unleaded)
Concentration or Composition (by Analyte or Components) 5000μg/mL
Chemical Name or Material Unleaded gasoline solution (unweathered)
CAS Max % 99.37
CAS 000067-56-1
CAS 008006-61-9
CAS Max % 0.63