Thermo Scientific™ Universal Piping for Thermo Scientific™ 1300 Series A2 Biological Safety Cabinets
Factory-installed Universal Air/Gas Piping
Supplier:  Thermo Scientific™ 1910286
 View more versions of this product
Catalog No. 13-261-352
Description
Description
Universal Piping for Air/Gas Service
  • Allows air and gas service to be piped through top, sides, or bottom of cabinet to minimize side-to-side footprint of cabinet utility piping
  • Two pre-plumbed service ports per side only
  • Factory-installed; must be ordered with cabinet
  • For cabinets with stainless-steel liners, specify using unique part number rather than ordering left and right side separately
Order Info
Factory installed. Must be specified when ordering cabinet.
Specifications
Specifications
Universal Piping
ft. models, right and top/bottom connection