Please login to your online account to display your discounted pricing

Universal Indicator, pH 2 - 10, Ricca Chemical


Quantity 20L
Packaging Cubitainer

Manufacturer: ricca chemical company  RIR887000020F

 View more versions of this product

Catalog No. R887000020F

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Color Clear dark
Quantity 20L
Chemical Name or Material Universal Indicator
CAS 64-17-5
CAS Max % 0.1
CAS 60-11-7
CAS Min % 0.07
CAS Max % 0.08
CAS 493-52-7
CAS Min % 0.05
CAS Max % 0.05
CAS 77-09-8
CAS Min % 0.02
CAS Max % 0.03
Name Note pH 2 - 10
CAS Min % 98.0
Packaging Cubitainer
CAS Max % 100.0
CAS 62625-21-2
CAS Min % 0.12
CAS Max % 0.13
CAS 34722-90-2
CAS Min % 0.1

Specifications

Chemical Name or Material Universal Indicator
Name Note pH 2 - 10
CAS 64-17-5
CAS Min % 98.0
CAS Max % 100.0
CAS 62625-21-2
CAS Min % 0.12
CAS Max % 0.13
CAS 34722-90-2
CAS Min % 0.1
CAS Max % 0.1
CAS 60-11-7
CAS Min % 0.07
CAS Max % 0.08
CAS 493-52-7
CAS Min % 0.05
CAS Max % 0.05
CAS 77-09-8
CAS Min % 0.02
CAS Max % 0.03
Physical Form Liquid