missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Universal Indicator, Bogens, Alcoholic, Ricca Chemical

$62.35 - $281.30

Chemical Identifiers

CAS 7732-18-5,67-63-0,62625-21-2,76-59-5,60-11-7,493-52-7
CAS Min % 63.0,34.57,0.06,0.05,0.04,0.03
CAS Max % 66.0,35.98,0.07,0.05,0.04,0.03
Physical Form Liquid
Chemical Name or Material Universal Indicator, Bogens, Alcoholic
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
887516 SDS Ricca Chemical Company
RI887516
Glass amber 500mL Each for $190.33
88754 Ricca Chemical Company
RI88754
Glass amber 120mL Each for $62.35
887532 SDS Ricca Chemical Company
RI887532
Glass amber 1L Each for $281.30
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,67-63-0,62625-21-2,76-59-5,60-11-7,493-52-7
66.0,35.98,0.07,0.05,0.04,0.03
Universal Indicator, Bogens, Alcoholic
63.0,34.57,0.06,0.05,0.04,0.03
Liquid

Specifications

7732-18-5,67-63-0,62625-21-2,76-59-5,60-11-7,493-52-7
66.0,35.98,0.07,0.05,0.04,0.03
Glass amber
500mL
63.0,34.57,0.06,0.05,0.04,0.03
Liquid
Clear light green
Universal Indicator, Bogens, Alcoholic