Upchurch Scientific™ Universal Column Couplers
$40.30 - $40.30
Specifications
Type Universal Column Coupler
For Use With (Application) Connect column w/internal 10-32 threads to precolumn or guard column w/10-32 threads
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With (Equipment) Swept Volume Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Swept Volume Price Quantity  
05-702-80 Upchurch Scientific™
UPU284
F-200 Derlin fingertights w/0.007 in. I.D. 1.3μL Each for $40.30
Specifications Description & Specifications
Specifications
Universal Column Coupler
Connect column w/internal 10-32 threads to precolumn or guard column w/10-32 threads