Please login to your online account to display your discounted pricing

Understanding Acids, Bases, and Salts


Manufacturer: fisher science education  AZ-20-1723

 View more versions of this product

Catalog No. S63469

  • / Each
 

Description & Specifications

Specifications

Class Size 40 students
Kit Size 40 students
Weight (English) 2 lb.
Weight (Metric) 0.9kg
For Use With (Application) Explore the chemical and physical properties of acids, bases, and salts, industrial applications
Product Type Chemistry Kit