missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Undecylenic aldehyde, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 157991000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Undecylenic aldehyde | |
112-45-8 | |
235°C | |
Authentic | |
Glass bottle | |
1.4417 to 1.4437 | |
MFCD00007032 | |
0.845 | |
Solubility in water: insoluble. Other solubilities: soluble in 80% alcohol,soluble in fixed oils,insoluble in glycerin | |
C=CCCCCCCCCC=O | |
168.28 | |
168.28 |
97% | |
0.8450g/mL | |
76°C | |
96% min. (GC) | |
C11H20O | |
H2C=CH(CH2)8CHO | |
100g | |
10-undecenal, undecylenic aldehyde, undecylenaldehyde, hendecenal, 10-undecen-1-al, 1-undecen-10-al, 10-undecylenealdehyde, 10-hendecenal, 10-undecenyl aldehyde, c-11 aldehyde, undecylenic | |
OFHHDSQXFXLTKC-UHFFFAOYSA-N | |
undec-10-enal | |
8187 | |
97% |
Safety and Handling
GHS H Statement
Causes skin irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
GHS Signal Word: Warning
EINECSNumber : 203-973-1
RTECSNumber : YQ2835000
TSCA : TSCA
RUO â Research Use Only