Please login to your online account to display your discounted pricing

Undecylenic Aldehyde 95%, Acros Organics


Quantity 100g

Manufacturer: thermo fisher scientific  AC388201000

 View more versions of this product

Catalog No. AC388201000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Formula Weight 168.28
Physical Form Liquid
Quantity 100g
Color Clear colorless
Density 0.845g/mL
CAS 112-45-8
Molecular Formula C11H20O
Refractive Index 1.4420 to 1.4450 (20°C, 589nm)
Synonym 10-Undecenal
Infrared Spectrum Authentic
Linear Formula H2C=CH(CH2)8CHO
Assay Percent Range 95%
Chemical Name or Material Undecylenic aldehyde
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.

Specifications

Chemical Name or Material Undecylenic aldehyde
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C11H20O
Linear Formula H2C=CH(CH2)8CHO
Formula Weight 168.28
Synonym 10-Undecenal
Accessories

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.

SDS & Safety Documents