Please login to your online account to display your discounted pricing

Alfa Aesar™ Undecanophenone, 97%

$42.47 - $167.78

Chemical Identifiers

Chemical Name or Material Undecanophenone
CAS 4433-30-1
Molecular Formula C17H26O
Formula Weight 246.39
MDL Number MFCD00051548
Beilstein 2211502
Synonym n-Decyl phenyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0211906

 
alfa aesar™
L0211906
5g Each for $42.47

AAL0211914

 
alfa aesar™
L0211914
25g Each for $167.78
Description & Specifications

Specifications

Formula Weight 246.39
Chemical Name or Material Undecanophenone
Assay 97%
Molecular Formula C17H26O
MDL Number MFCD00051548
CAS 4433-30-1
Melting Point 28° to 30°C
Beilstein 2211502
Synonym n-Decyl phenyl ketone
Boiling Point 202° to 204°C (14mmHg)
Quantity 5g