missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Undecanoic acid, 98%

Manufacturer:  Alfa Aesar™ A112440B

 View more versions of this product

Catalog No. AAA112440B


 

Description

Description

Applications
Undecanoic acid is used as an unsaturated fatty acid. It is also used as intermediates of Liquid Crystals. It is used in organic synthesis, Catalytic agent, Petrochemical additive. It is also used in anti-fungal agent, perfumery chemicals.

Solubility
Insoluble in water.

Notes
Store at 4°C. Incompatible with bases, oxidizing agents, reducing agents. Keep container closed when not in use.
Specifications

Specifications

Undecanoic acid
98%
MFCD00002730
ZDPHROOEEOARMN-UHFFFAOYSA-N
undecanoic acid
8180
186.3
283°C to 285°C
>110°C (230°F)
112-37-8
C11H22O2
1759287
CCCCCCCCCCC(=O)O
186.295
CHEBI:32368
26°C to 29°C
1000g

Chemical Identifiers

112-37-8
186.295
ZDPHROOEEOARMN-UHFFFAOYSA-N
CHEBI:32368
C11H22O2
MFCD00002730
8180
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : YQ2275000

TSCA : TSCA

SDS Documents