missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Undecane-1,11-dicarboxylic acid, 95%

$24.05 - $93.92

Chemical Identifiers

CAS 505-52-2
Molecular Formula C13H24O4
Molecular Weight (g/mol) 244.331
MDL Number MFCD00002740
InChI Key DXNCZXXFRKPEPY-UHFFFAOYSA-N
Synonym brassylic acid, 1,11-undecanedicarboxylic acid, brassilic acid, 1,13-tridecanedioic acid, brassylate, unii-pl3iq40c34, undecane-1,11-dicarboxylic acid, tridecanedioate, dsstox_cid_1683, dsstox_rid_76281
PubChem CID 10458
ChEBI CHEBI:73718
IUPAC Name tridecanedioic acid
SMILES C(CCCCCC(=O)O)CCCCCC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6096203 View Documents Alfa Aesar™
H6096203
1g Each for $24.05
 
AAH6096206 View Documents Alfa Aesar™
H6096206
5g Each for $93.92
 
Specifications

Chemical Identifiers

505-52-2
244.331
DXNCZXXFRKPEPY-UHFFFAOYSA-N
10458
tridecanedioic acid
C13H24O4
MFCD00002740
brassylic acid, 1,11-undecanedicarboxylic acid, brassilic acid, 1,13-tridecanedioic acid, brassylate, unii-pl3iq40c34, undecane-1,11-dicarboxylic acid, tridecanedioate, dsstox_cid_1683, dsstox_rid_76281
CHEBI:73718
C(CCCCCC(=O)O)CCCCCC(=O)O

Specifications

1g
C13H24O4
MFCD00002740
DXNCZXXFRKPEPY-UHFFFAOYSA-N
tridecanedioic acid
10458
244.33
Undecane-1,11-dicarboxylic acid
505-52-2
HO2C(CH2)11CO2H
brassylic acid, 1,11-undecanedicarboxylic acid, brassilic acid, 1,13-tridecanedioic acid, brassylate, unii-pl3iq40c34, undecane-1,11-dicarboxylic acid, tridecanedioate, dsstox_cid_1683, dsstox_rid_76281
C(CCCCCC(=O)O)CCCCCC(=O)O
244.331
CHEBI:73718
95%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA