Learn More
Undecanal, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1610114
Description
Undecanal is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUndecanal is used as a pharmaceutical intermediate.
Solubility
Soluble in dipropylene glycol, fixed oils, propylene glycol and water (14.27 mg/L at 25°C)
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents, reducing agents and bases. It is sensitive to air.
Specifications
Undecanal | |
-2°C | |
110°C to 112°C (12 mmHg) | |
C11H22O | |
CH3(CH2)9CHO | |
25 g | |
Air Sensitive | |
Soluble in dipropylene glycol,fixed oils,propylene glycol and water (14.27mg/L at 25°C) | |
CCCCCCCCCCC=O | |
170.296 | |
CHEBI:46202 | |
97% |
112-44-7 | |
0.827 | |
95°C (203°F) | |
1.434 | |
MFCD00007016 | |
1753213 | |
undecanaldehyde, n-undecanal, undecyl aldehyde, undecylic aldehyde, 1-undecanal, undecylaldehyde, hendecanal, hendecanaldehyde, aldehyde c-11, n-undecyl aldehyde | |
KMPQYAYAQWNLME-UHFFFAOYSA-N | |
undecanal | |
8186 | |
170.3 |
Chemical Identifiers
112-44-7 | |
170.296 | |
KMPQYAYAQWNLME-UHFFFAOYSA-N | |
8186 | |
undecanal |
C11H22O | |
MFCD00007016 | |
undecanaldehyde, n-undecanal, undecyl aldehyde, undecylic aldehyde, 1-undecanal, undecylaldehyde, hendecanal, hendecanaldehyde, aldehyde c-11, n-undecyl aldehyde | |
CHEBI:46202 | |
CCCCCCCCCCC=O |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 203-972-6
RTECSNumber : YQ1500000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only