missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
γ-Undecalactone, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC259495000
Specifications
γ-Undecalactone | |
145°C | |
Glass bottle | |
0.9425 | |
Colorless | |
104-67-6 | |
C11H20O2 | |
1,4-undecanolide, 2 3h-furanone, 5-heptyldihydro, 4-undecanolide, 5-heptyldihydrofuran-2 3h-one, aldehyde c-14 peach, gamma-undecalactone, gamma-undecanolactone, peach aldehyde, peach lactone, persicol | |
CCCCCCCC1CCC(=O)O1 | |
184.28 | |
184.28 | |
98% |
0.9425g/mL | |
Authentic | |
1.4490 to 1.4540 | |
286.0°C | |
500mL | |
97.5% min. (GC) | |
MFCD00005405 | |
PHXATPHONSXBIL-UHFFFAOYSA-N | |
5-heptyloxolan-2-one | |
7714 | |
Liquid |
Chemical Identifiers
104-67-6 | |
184.28 | |
PHXATPHONSXBIL-UHFFFAOYSA-N | |
7714 | |
CCCCCCCC1CCC(=O)O1 |
C11H20O2 | |
MFCD00005405 | |
1,4-undecanolide, 2 3h-furanone, 5-heptyldihydro, 4-undecanolide, 5-heptyldihydrofuran-2 3h-one, aldehyde c-14 peach, gamma-undecalactone, gamma-undecanolactone, peach aldehyde, peach lactone, persicol | |
5-heptyloxolan-2-one |
Safety and Handling
GHS H Statement
Harmful to aquatic life with long lasting effects.
GHS P Statement
Avoid release to the environment.
Dispose of contents/container to an approved waste disposal plant.
EINECSNumber : 203-225-4