missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

γ-Undecalactone, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC259491000

 View more versions of this product

Catalog No. AC259491000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

γ-Undecalactone
145°C
Glass bottle
0.9425
Colorless
104-67-6
C11H20O2
1,4-undecanolide, 2 3h-furanone, 5-heptyldihydro, 4-undecanolide, 5-heptyldihydrofuran-2 3h-one, aldehyde c-14 peach, gamma-undecalactone, gamma-undecanolactone, peach aldehyde, peach lactone, persicol
CCCCCCCC1CCC(=O)O1
184.28
184.28
98%
0.9425g/mL
Authentic
1.4490 to 1.4540
286.0°C
100mL
97.5% min. (GC)
MFCD00005405
PHXATPHONSXBIL-UHFFFAOYSA-N
5-heptyloxolan-2-one
7714
Liquid

Chemical Identifiers

104-67-6
184.28
PHXATPHONSXBIL-UHFFFAOYSA-N
7714
CCCCCCCC1CCC(=O)O1
C11H20O2
MFCD00005405
1,4-undecanolide, 2 3h-furanone, 5-heptyldihydro, 4-undecanolide, 5-heptyldihydrofuran-2 3h-one, aldehyde c-14 peach, gamma-undecalactone, gamma-undecanolactone, peach aldehyde, peach lactone, persicol
5-heptyloxolan-2-one
Safety and Handling

Safety and Handling

GHS H Statement
Harmful to aquatic life with long lasting effects.

GHS P Statement
Avoid release to the environment.
Dispose of contents/container to an approved waste disposal plant.

EINECSNumber : 203-225-4

SDS Documents