Please login to your online account to display your discounted pricing

UNC 926 hydrochloride, Tocris™

L3MBTL1 domain inhibitor

$218.74 - $915.10

Chemical Identifiers

Chemical Name or Material UNC 926 hydrochloride
Assay Percent Range >99%
Molecular Formula C16H21BrN2O·HCl
Formula Weight 373.72
Synonym (3-Bromophenyl)[4-(1-prrolidinyl)-1-piperidinyl]methanone hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

451610

 
r & d systems
RDS451610
10mg Each for $218.74

451650

 
r & d systems
RDS451650
50mg Each for $915.10
Description & Specifications

Specifications

Formula Weight 373.72
Quantity 10mg
Synonym (3-Bromophenyl)[4-(1-prrolidinyl)-1-piperidinyl]methanone hydrochloride
Chemical Name or Material UNC 926 hydrochloride
Assay Percent Range >99%
Molecular Formula C16H21BrN2O·HCl
CAS 1184136-10-4
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature