Please login to your online account to display your discounted pricing

UNC 0646, Tocris™

Potent and selective G9a/GLP inhibitor

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material UNC 0646
Assay Percent Range >99%
Molecular Formula C36H59N7O2
Formula Weight 621.9
Synonym N-(1-Cyclohexyl-4-piperidinyl)-2-[hexahydro-4-(1-methylethyl)-1H-1, 4-diazepin-1-yl]-6-methoxy-7-[3-(1-piperidinyl)propoxy]-4-quinazolinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

434250

 
r & d systems
434250
50mg Each for $1,050.42

434210

 
r & d systems
434210
10mg Each for $252.39
Description & Specifications

Specifications

Formula Weight 621.9
Quantity 50mg
CAS 1320288-17-2
Recommended Storage Store at +4°C
Chemical Name or Material UNC 0646
Assay Percent Range >99%
Molecular Formula C36H59N7O2
Synonym N-(1-Cyclohexyl-4-piperidinyl)-2-[hexahydro-4-(1-methylethyl)-1H-1, 4-diazepin-1-yl]-6-methoxy-7-[3-(1-piperidinyl)propoxy]-4-quinazolinamine
Safety and Handling

Store at +4°C