Please login to your online account to display your discounted pricing

UNC 0224, Tocris™

Potent G9a histone lysine methyltransferase inhibitor

$264.88 - $1,099.17

Chemical Identifiers

Chemical Name or Material UNC 0224
Assay Percent Range >98%
Molecular Formula C26H43N7O2
Formula Weight 485.67
Synonym 7-[3-(Dimethylamino)propoxy]-2-(hexahydro-4-methyl-1H-1, 4-diazepin-1-yl)-6-methoxy-N-(1-methyl-4-piperidinyl)-4-quinazolinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

386110

 
r & d systems
386110
10mg Each for $264.88

386150

 
r & d systems
386150
50mg Each for $1,099.17
Description & Specifications

Specifications

Formula Weight 485.67
Quantity 10mg
Chemical Name or Material UNC 0224
Assay Percent Range >98%
Molecular Formula C26H43N7O2
CAS 1197196-48-7
Synonym 7-[3-(Dimethylamino)propoxy]-2-(hexahydro-4-methyl-1H-1, 4-diazepin-1-yl)-6-methoxy-N-(1-methyl-4-piperidinyl)-4-quinazolinamine
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C