Please login to your online account to display your discounted pricing

Thermo Scientific™ Unassembled Convenience Kits: EPA Vials with Caps and Septa


Designed for discrete water sampling applications.

Manufacturer: thermo scientific™  B7920B

 View more versions of this product

Catalog No. 03-377-32

  • / Pack of 100 Each
 

Description & Specifications

Specifications

For Use With discrete water sampling applications
Item Description Clear; 20mL
Type Unassembled EPA Vial Kit
Volume (Metric) 20mL
Color Clear
Description Clear 20mL vials, PTFE/Silicone septa and cap

Thermo Scientific™ National EPA Vial Convenience Kits are designed for discrete water sampling applications.

Each carton contains shrink-wrapped vials and separately packaged caps and natural-colored PTFE/clear Silicone septa in polybags.