Ultraspec™ 2000 and 2001 Eyewear
Adjustable side temple for a customized fit.
Manufacturer:  Fisher Science Education S0250XEMD
 View more versions of this product
Catalog No. S43186
Description
Description
  • Frameless lens features Ultradura™ hardcoat, scratch-resistant coating
  • S43187 has a larger lens to fit over most glasses and Duoflex™ comfort cushions at ends of the temples
Compliance
Meets ANSI™ Z87.1-1989 standards for impact protection.
Specifications
Specifications
Meet ANSI Z87.1-1989 standards for impact protection
Spatula
Anti-fog,Scratch-resistant
Clear
Clear
Education
Documents