Branson Ultrasonics™ Ultrasonic Industrial Cleaning Solutions
Safely clean metals including stainless-steel, tin, and copper
Manufacturer:  Branson Ultrasonics™ 000955114
 View more versions of this product
Catalog No. 15-345-113
Description
Description
Industrial-strength.
  • Safely and effectively remove soils, rust, and oxides from a variety of materials
  • Solutions contain no phosphates and are biodegradable
  • Dilute with water to the recommended working concentration listed on ordering table
Specifications
Specifications
Cutting Oils, Metal Carbon
1 qt.
8 to 10%
11 to 12
Documents