Branson Ultrasonics™ Ultrasonic Cleaning Oxide Removers
Safely clean metals including stainless-steel, tin, and copper
Manufacturer:  Branson Ultrasonics™ 000955516
 View more versions of this product
Catalog No. 15-335-46
Description
Description
  • Safely and effectively remove soils, rust, and oxides from a variety of materials
  • Solutions contain no phosphates and are biodegradable
  • Dilute with water to the recommended working concentration listed on ordering table
Specifications
Specifications
Rust, Metal Oxides
6 to 7%
Oxide Remover
1 gal.
3 to 4
Documents