Please login to your online account to display your discounted pricing

Alfa Aesar™ Ultramark™ 2500F

$128.82 - $128.82

Chemical Identifiers

Chemical Name or Material Ultramark™ 2500F
CAS 66795-94-6
MDL Number MFCD00166131
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1669603

  SDS
alfa aesar™
L1669603
1g Each for $128.82
Description & Specifications

Specifications

Chemical Name or Material Ultramark™ 2500F
MDL Number MFCD00166131
Boiling Point 183° to 210°C (0.7mmHg)
Quantity 1g
CAS 66795-94-6