Please login to your online account to display your discounted pricing

Alfa Aesar™ Ultramark™ 2500F


Quantity 1g

Manufacturer: alfa aesar™  L1669603

 View more versions of this product

Catalog No. AAL1669603

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Ultramark™ 2500F
MDL Number MFCD00166131
Quantity 1g
CAS 66795-94-6
Boiling Point 183° to 210°C (0.7mmHg)

Specifications

Chemical Name or Material Ultramark™ 2500F
CAS 66795-94-6
MDL Number MFCD00166131