Please login to your online account to display your discounted pricing

Alfa Aesar™ Ultramark™ 1960F

$113.62 - $113.62

Chemical Identifiers

Chemical Name or Material Ultramark™ 1960F
CAS 66795-94-6
MDL Number MFCD00166131
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1669503

  SDS
alfa aesar™
L1669503
1g Each for $113.62
Description & Specifications

Specifications

Quantity 1g
CAS 66795-94-6
MDL Number MFCD00166131
Boiling Point 128°C (0.2mmHg)
Chemical Name or Material Ultramark™ 1960F