Please login to your online account to display your discounted pricing

Alfa Aesar™ Ultramark™ 1960F


Quantity 1g

Manufacturer: alfa aesar™  L1669503

 View more versions of this product

Catalog No. AAL1669503

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 1g
CAS 66795-94-6
MDL Number MFCD00166131
Boiling Point 128°C (0.2mmHg)
Chemical Name or Material Ultramark™ 1960F

Specifications

Chemical Name or Material Ultramark™ 1960F
CAS 66795-94-6
MDL Number MFCD00166131