MilliporeSigma™ Ultrafiltration Membranes - Amicon™ XM Polyacrylonitrile/PVC Copolymer Filters
High-recovery membranes for stirred cells
$185.00 - $413.00
Specifications
Material Polyacrylonitrile, PVC Co-polymer
For Use With (Application) High recovery membranes for stirred cells
Quantity 10 Pack
Type Ultrafiltration Disc
View More Specs
Products 3
Catalog Number Mfr. No. Diameter (English) Diameter (Metric) MWCO Price Quantity  
Catalog Number Mfr. No. Diameter (English) Diameter (Metric) MWCO Price Quantity  
14112AM
View Documents Promotion Details
MilliporeSigma™
14112AM
0.98 in. 2.5 cm 50,000 NMWL
Pack of 10 Each for $185.00
14322
View Documents Promotion Details
MilliporeSigma™
14322
1.75 in. 4.45 cm 300,000 NMWL
Pack of 10 Each for $319.00
14342
View Documents Promotion Details
MilliporeSigma™
14342
2.99 in. 7.6 cm 300,000 NMWL
Pack of 10 Each for $413.00
Description
Description
  • Regenerated cellulose membranes
  • For concentration of hydrophobic samples or to desalt extremely dilute solutions
  • Hydrophilic, tight microstructure assures high retention with low adsorption of protein, DNA, or other macromolecules
†43mm nominal diameter.
‡62mm nominal diameter.
Specifications
Specifications
Polyacrylonitrile, PVC Co-polymer
High recovery membranes for stirred cells
10 Pack
Ultrafiltration Disc