Please login to your online account to display your discounted pricing

Thermo Scientific™ UltraClean™ Microarray Slides

Expand your research options with Thermo Scientific™ UltraClean™ Microarray Slides, for use in any coating process compatible with glass.

$391.93 - $547.51

Specifications

Description UltraClean Microarray Slides
Length (Metric) 75mm
Thickness (Metric) 1mm
Width (Metric) 25mm
View More Specs
Products
Catalog Number Mfr. No. Enhanced Surface Price Quantity    

22-037-213

 
thermo scientific™
ERC225128M20
No Pack of 20 Each for $391.93

22-037-215

 
thermo fisher scientific
ERC245587M20
Yes Pack of 20 Each for $547.51
Description & Specifications

Specifications

Description UltraClean Microarray Slides
Length (Metric) 75mm
Thickness (Metric) 1mm
Width (Metric) 25mm

Expand your research options with Thermo Scientific™ UltraClean™ Microarray Slides, for use in any coating process compatible with glass - including silanization, other functionalization such as Poly-Lysine, or layering of thin films.  These slides are compatible with standard microarray spotting and detection systems.

Features:

  • Manufactured from high-quality, ultra-flat, low-fluorescence glass.
  • Packaged in a secure, no-contact mBox™ and vacuum sealed in a foil barrier bag
Overall Dimensions:
  • 25 x 75mm