missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

UK 78282 hydrochloride, Tocris™

Blocker of KV1.3 and KV1.4 channels

$250.58 - $1,042.86

Chemical Identifiers

Assay Percent Range >99%
Molecular Formula C29H35NO2·HCl
Formula Weight 466.05
Synonym 4-[(Diphenylmethoxy)methyl]-1-[3-(4-methoxyphenyl)propyl]-piperidine hydrochloride
Chemical Name or Material UK 78282 hydrochloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
382910 R & D Systems
RDS382910
10mg Each for $250.58
382950 R & D Systems
RDS382950
50mg Each for $1,042.86
Specifications Description & Specifications

Chemical Identifiers

>99%
466.05
UK 78282 hydrochloride
C29H35NO2·HCl
4-[(Diphenylmethoxy)methyl]-1-[3-(4-methoxyphenyl)propyl]-piperidine hydrochloride

Specifications

136647-02-4
C29H35NO2·HCl
4-[(Diphenylmethoxy)methyl]-1-[3-(4-methoxyphenyl)propyl]-piperidine hydrochloride
UK 78282 hydrochloride
>99%
466.05
10mg

Safety and Handling

Recommended Storage : Store at -20°C