Please login to your online account to display your discounted pricing

UK 14,304, Tocris

α2 agonist

$149.90 - $632.23

Chemical Identifiers

Chemical Name or Material UK 14, 304
Assay Percent Range >98%
Molecular Formula C11H10BrN5
Formula Weight 292.14
Synonym 5-Bromo-6-(2-imidazolin-2-ylamino)quinoxaline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

042550

 
r & d systems
RDS042550
50mg Each for $632.23

042510

 
r & d systems
RDS042510
10mg Each for $149.90
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 292.14
Molecular Formula C11H10BrN5
Chemical Name or Material UK 14, 304
Assay Percent Range >98%
CAS 59803-98-4
Synonym 5-Bromo-6-(2-imidazolin-2-ylamino)quinoxaline
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature