Please login to your online account to display your discounted pricing

(+)-UH 232 maleate, Tocris


D2 autoreceptor antagonist; Also D3 partial agonist

Quantity 50mg

Manufacturer: r & d systems  RDS077550

 View more versions of this product

Catalog No. 077550

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 391.51
Chemical Name or Material (+)-UH 232 maleate
Assay Percent Range >98%
Molecular Formula C18H29NO·C4H4O4
CAS 1217473-50-1
Synonym cis-(+)-5-Methoxy-1-methyl-2-(di-N-propylamino)tetralin maleate
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material (+)-UH 232 maleate
Assay Percent Range >98%
Molecular Formula C18H29NO·C4H4O4
Formula Weight 391.51
Synonym cis-(+)-5-Methoxy-1-methyl-2-(di-N-propylamino)tetralin maleate
Accessories

Store at Room Temperature