Please login to your online account to display your discounted pricing

UCM 707, Tocris

Potent anandamide transport inhibitor

$221.01 - $904.70

Chemical Identifiers

Chemical Name or Material UCM 707
Assay Percent Range >98%
Molecular Formula C25H37NO2
Formula Weight 383.57
Synonym (5Z,8Z,11Z,14Z)-N-(3-Furanylmethyl)-5,8,11,14-eicosatetraenamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

196610

 
r & d systems
RDS196610
10mg Each for $221.01

196650

 
r & d systems
RDS196650
50mg Each for $904.70
Description & Specifications

Specifications

Formula Weight 383.57
Quantity 10mg
Molecular Formula C25H37NO2
Synonym (5Z,8Z,11Z,14Z)-N-(3-Furanylmethyl)-5,8,11,14-eicosatetraenamide
Chemical Name or Material UCM 707
Assay Percent Range >98%
CAS 390824-20-1
Recommended Storage Desiccate at -20°C
Safety and Handling

Desiccate at -20°C