Please login to your online account to display your discounted pricing

UCL 2077, Tocris

Slow afterhyperpolarization (sAHP) channel blocker

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material UCL 2077
Assay Percent Range >99%
Molecular Formula C25H22N2
Formula Weight 350.46
Synonym N-Trityl-3-pyridinemethanamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

266110

 
r & d systems
RDS266110
10mg Each for $177.37

266150

 
r & d systems
RDS266150
50mg Each for $746.48
Description & Specifications

Specifications

Formula Weight 350.46
Quantity 10mg
Chemical Name or Material UCL 2077
Assay Percent Range >99%
Molecular Formula C25H22N2
CAS 918311-87-2
Synonym N-Trityl-3-pyridinemethanamine
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature